Sodium chloride, Sigma-Aldrich®
Catalog # SIAL07982-100TAB-F
Supplier: Merck
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477