TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®
Catalog # SIALT9269-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: TAPSO, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid
:
:M-Clarity™ quality level = MQ200
:259.28 g/mol
EINECS:269-993-8
CAS Number:68399-81-5
Formula:C₇H₁₇NO₇S
MDL Number:MFCD00038352