Order Entry
Slovenia
ContactUsLinkComponent
Organic reference standard, Dimethyl phthalate 5,000 µg/ml in methanol
Organic reference standard, Dimethyl phthalate 5,000 µg/ml in methanol
Catalog # PROOU-EPA-1112
CAS Number:  
Organic reference standard, Dimethyl phthalate 5,000 µg/ml in methanol
Catalog # PROOU-EPA-1112
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    131-11-3
  • Description:
    Dimethyl phthalate 5,000 µg/ml in methanol
  • Formula:
    C₁₀H₁₀O₄
  • Description:
    Dimethyl phthalate

Specifications

About this item