Orders LinkContactUsLinkComponent
Multi purpose bottles
Multi purpose bottles
Supplier:   RATIOLAB
Multi purpose bottles
Supplier:  RATIOLAB
Certificates
Multi purpose bottles
Multi purpose bottles
3 Options Available Below
Product Details & Documents
Polyethylene, blue translucent.
  • Incl. lid with nonwoven fabric liner
  • With graduations
  • Light protection
  • Wide neck diameter of 80 mm
Product Family Options
Product Information
  • Capacity
    Colour
    Neck-Ø
    Availability
    Price
  • RATI5922250
    Multipurpose collecting bottles with label
    Multipurpose collecting bottles with label
    2500 ml
    Blue
    80 mm
    Log in for availability
    Log in for availability
    1 * 60 items
    1 * 60 items - 171,98 €
    171,98 €
    1 * 60 items
       171,98 €
  • RATI5922204
    Lids for multipurpose collecting bottles, 2000 or 2500 ml capacity
    Lids for multipurpose collecting bottles, 2000 or 2500 ml capacity
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • RATI5922253
    Labels, self-adhesive, for multi purpose bottles
    Labels, self-adhesive, for multi purpose bottles
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items