Orders LinkContactUsLinkComponent
Bossheads for frames, 90°
 :   BOCHEM
Bossheads for frames, 90°
 :  BOCHEM
Bossheads for frames, 90°
Bossheads for frames, 90°
Bossheads for frames, either made of stainless steel or malleable cast iron, chromium plated.
  • For rods up to 13 mm
  • Angle: 90°
  • With socket screw (M6)
Product Family Options
  • Material
    For rod Ø
  • Tempered cast iron
    13 mm
    1 * 1 items
    1 * 1 items - 11,00 €
    11,00 €
    1 * 1 items
       11,00 €
  • Stainless steel 18/10
    13 mm
    1 * 1 items
    1 * 1 items - 45,30 €
    45,30 €
    1 * 1 items
       45,30 €