Orders LinkContactUsLinkComponent
Bossheads with hook
Bossheads with hook
Supplier:   BOCHEM
Bossheads with hook
Supplier:  BOCHEM
Certificates
Bossheads with hook
Bossheads with hook
2 Options Available Below
Product Details & Documents
Bossheads with hook, either made of stainless steel or nickel plated steel.
  • For rods up to 13 mm
  • Angle: 90°
Product Family Options
Product Information
  • Material
    For rod Ø
    Availability
    Price
  • Stainless steel 18/10, anti-magnetic
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 23,30 €
    23,30 €
    1 * 1 items
       23,30 €
  • Steel, nickel plated
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 11,40 €
    11,40 €
    1 * 1 items
       11,40 €