Orders LinkContactUsLinkComponent
Sample containers, Pure
Sample containers, Pure
Supplier:   BURKLE
Sample containers, Pure
Supplier:  BURKLE
Certificates
Sample containers, Pure
Sample containers, Pure
3 Options Available Below
Product Details & Documents
Sample box with tightly closing sealing cover.
  • Tightly closing sample container
  • With attached, vertically fixable snap-on lid
  • PP, transparent
The lid can be opened with one hand and its hinge angle is 90°. Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, and nothing can drip. The box is completely tight, has no crevices, and is therefore easy to fill and can be emptied without residues. It is autoclavable and suitable for deep freezing.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • BURK6206-0100
    Graduated to 80 ml
    Graduated to 80 ml
    90 ml
    43 mm
    80 mm
    Log in for availability
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,60 €
    0,60 €
    Log in for availability
    1 * 350 items
    1 * 350 items - 129,50 €
    129,50 €
    1 * 1 items
       0,60 €
    1 * 350 items
       129,50 €
    Specifications:
    New Product
    New Product
  • BURK6206-0060
    With labeling area and graduation
    With labeling area and graduation
    50 ml
    31 mm
    78 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 0,50 €
    0,50 €
    1 * 1 items
       0,50 €
  • Graduated
    300 ml
    75 mm
    88 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1,40 €
    1,40 €
    1 * 1 items
       1,40 €