Orders LinkContactUsLinkComponent
Containers with screw cap, multipurpose
Containers with screw cap, multipurpose
Supplier:   BURKLE
Containers with screw cap, multipurpose
Supplier:  BURKLE
Certificates
Containers with screw cap, multipurpose
Containers with screw cap, multipurpose
3 Options Available Below
Product Details & Documents
PP, translucent, with PE screw lid.
  • Suitable for use with foodstuffs
  • No crevices
  • Very good chemical and temperature resistance
Contents can be recognised easily due to the transparent sides. Emptying without residues.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Colour
    Availability
    Price
  • 1000 ml
    126 mm
    126 mm
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 12,00 €
    12,00 €
    1 * 1 items
       12,00 €
  • 250 ml
    104 mm
    50 mm
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 5,20 €
    5,20 €
    1 * 1 items
       5,20 €
  • 500 ml
    104 mm
    100 mm
    Transparent
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6,80 €
    6,80 €
    1 * 1 items
       6,80 €