ContactUsLinkComponent
Condenser clamps, swivel-type
Catalog # 241-7425
Supplier:  BOCHEM
undefined
Condenser clamps, swivel-type
Catalog # 241-7425
Supplier:  BOCHEM
Specifications
  • For:
    Condensers
  • Spanning width:
    50 - 100 mm
  • Length:
    150 mm
  • Material:
    Cast iron
  • Pk:
    1
Specifications
About this item
Malleable cast iron.
  • Swivel type
  • Chromium plated
  • Silicone coated