Orders LinkContactUsLinkComponent
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
Filter vials, SINGLE StEP™
Simple replacements for HPLC vials, HPLC caps, syringes, needles, and syringe filters. Filter vials minimise steps and loss of product. They protect HPLC, GC, GC-MS and LC-MS systems against contaminants and particulates.
  • Compatible with both UHPLC and standard HPLC autosamplers
  • Colour coded pre-slit caps for easy identification of membrane and porosity
  • Low dead volume unit contains rugged PP insert with 450 µl loading capacity
Product Family Options
  • Membrane
    Pore size
    Colour
  • Nylon
    0,2 µm
    Black
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • Nylon
    0,2 µm
    Black
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €
  • PES
    0,2 µm
    Grey
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • PES
    0,2 µm
    Grey
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €
  • PTFE
    0,2 µm
    Green
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • PTFE
    0,2 µm
    Green
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €
  • PVDF
    0,2 µm
    Red
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • PVDF
    0,2 µm
    Red
    1 * 100 items
    1 * 100 items - 1 860,00 €
    1 860,00 €
    1 * 100 items
       1 860,00 €
  • Nylon
    0,45 µm
    Pink
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • Nylon
    0,45 µm
    Pink
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €
  • PTFE
    0,45 µm
    Blue
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • PTFE
    0,45 µm
    Blue
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €
  • PVDF
    0,45 µm
    Yellow
    1 * 200 items
    1 * 200 items - 742,00 €
    742,00 €
    1 * 200 items
       742,00 €
  • PVDF
    0,45 µm
    Yellow
    1 * 500 items
    1 * 500 items - 1 860,00 €
    1 860,00 €
    1 * 500 items
       1 860,00 €