ContactUsLinkComponent
Round slide mailer, LockMailer™
Round slide mailer, LockMailer™
Catalog # SIMPM950-4MA
Supplier:  Simport Scientific
undefined
Round slide mailer, LockMailer™
Catalog # SIMPM950-4MA
Supplier:  Simport Scientific
Specifications
  • No. of slides:
    4
  • Colour:
    Magenta
  • Description:
    Slide containers
  • Material:
    HDPE
  • Pk:
    500
Specifications
About this item
Transparent container made of high density polyethylene for mailing or storing slides between the doctor's office and the lab.
  • Inside channels slotted to keep slides separated
  • Unique tamper evident leakproof screw cap
  • Good stability on a bench