Orders LinkContactUsLinkComponent
Digital timer
Digital timer
Electronic timer, White, 50×23×62 mm
Catalog # KART902
Supplier:   KARTELL
Digital timer
Catalog # KART902
Supplier:  KARTELL
Digital timer
Digital timer
Electronic timer, White, 50×23×62 mm
Price
55,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Electronic timer, battery operated.
  • Four digits display up to 19 hours 59 minutes
  • Alarm sounds at the end of the set time
  • Free-standing, clip or magnetic fixings
Specifications
  • Colour:
    White
  • Description:
    Electronic timer
  • W×D×H:
    50×23×62 mm