ContactUsLinkComponent
Microbalance, Excellence XS3DU
Microbalance, Excellence XS3DU
  611-2454
Microbalance, Excellence XS3DU
  611-2454
  • Model:
    XS3DU
  • Pk:
    1
  • Readability:
    1 / 10 µg
  • Weighing cap.:
    0,8 / 3,1 g
  • Repeatability:
    0,8±* / 5±* µg
An economical microbalance that delivers reliable measurement performance for straightforward weighing of microgramme samples.
  • Monochrome touch screen display for simple intuitive control
  • FACT fully automatic time and temperature controlled internal adjustment
  • SmartTrac to provide a graphical track of capacity and weighing tolerances
  • Manually driven draught shield
  • Slot for second interface to provide range of connectivity options
The five pre-programmed applications and three user-definable fields for sample and operator identification provide sufficient data for simple weighing tasks.
 : For details of the full range of accessories, including options for filter weighing and antistatic options, please contact your local Avantor sales office.
 : * Measured at low load conditions (0,2g)