Orders LinkContactUsLinkComponent
Tanks
Tanks
Supplier:   KARTELL
Tanks
Supplier:  KARTELL
Certificates
Tanks
3 Options Available Below
Product Details & Documents
Made of HDPE, grey.
  • Moulded grips
  • Sturdy
Useful general purpose tanks that can be stacked for storage.
Product Family Options
Product Information
  • Colour
    Capacity
    W×D×H
    Availability
    Price
  • Grey
    27 l
    340×455×250 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 70,10 €
    70,10 €
    1 * 1 items
       70,10 €
  • Grey
    40 l
    350×560×304 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 97,80 €
    97,80 €
    1 * 1 items
       97,80 €
  • Grey
    75 l
    457×750×310 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 168,00 €
    168,00 €
    1 * 1 items
       168,00 €