2,3,4,5-Tetrachlorophenol
: CHEMPUR
- :4901-51-3
- :C₆H₂Cl₄O
- :231.89 g/mol
- :289 °C (1013 hPa)
- :116 °C
- :1.6 g/cm³ (20 °C)
- :225-531-7
- :2020
- :6.1,III
- :Density: 1.6 g/cm³ (20 °C)
- :4901-51-3
- :C₆H₂Cl₄O
- :231.89 g/mol
- :289 °C (1013 hPa)
- :116 °C
- :1.6 g/cm³ (20 °C)
- :225-531-7
- :2020
- :6.1,III
- :Density: 1.6 g/cm³ (20 °C)
Product Family Options
- Pack typePk
This product has been blocked by your organization. Please contact your purchasing department for more information