2,4-Dihydroxybenzoic acid
Catalog # MOLEM43391076
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:154.12 g/mol
Merck Index:12,08325
Boiling Pt:390 °C (1013 hPa)
Melting Pt:218…220 °C
Density:1.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-946-9
CAS Number:89-86-1
Formula:C₇H₆O₄
MDL Number:MFCD00002451