Orders LinkContactUsLinkComponent
BD® Blunt Needles
BD® Blunt Needles
BD® Blunt Needles
BD® Blunt Needles
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
  • The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
  • The blunt filter is designed for safe ampoule medication preparation
  • BD blunt fill needles have female luer fittings, which mate to male luer fittings
  • BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
  • Natural rubber latex is not part of the material formulation
  • Sterile, singe use
Caution: Note: The BD blunt fill and filter needles are not for skin injection.
Product Family Options
Blunt filter needle
  • Gauge
    Length
    Type of tip
    Type
    Colour code
  • 18 G
    40 mm
    Blunt bevel
    Sterile
    Lilac
    1 * 100 items
    1 * 100 items - 95,30 €
    95,30 €
    1 * 100 items
       95,30 €
Blunt fill needle
  • Gauge
    Length
    Type of tip
    Type
    Colour code
  • 18 G
    25 mm
    Blunt bevel
    Sterile
    Red
    1 * 100 items
    1 * 100 items - 72,40 €
    72,40 €
    1 * 100 items
       72,40 €
  • 18 G
    40 mm
    Blunt bevel
    Sterile
    Red
    1 * 100 items
    1 * 100 items - 27,50 €
    27,50 €
    1 * 100 items
       27,50 €
  • 18 G
    40 mm
    Blunt bevel
    Sterile
    Red