Orders LinkContactUsLinkComponent
Buckets, Square, PE
Supplier:   BURKLE
Buckets, Square, PE
Supplier:  BURKLE
Certificates
Buckets, Square, PE
Buckets, Square, PE
2 Options Available Below
Product Details & Documents
Made of PE, transparent.
  • Graduation 500 ml, up to 12 L
  • Saves space
  • Sturdy, easy to carry
  • Contents right to the top 14 L
  • Lid must be ordered seperately
Bucket for transporting bulky goods.
Product Family Options
Lids for square buckets
Product Information
  • Colour
    Capacity
    Height
    Availability
    Price
  • BURK2350-0141
    Lid for square buckets
    Lid for square buckets
    14 l
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 13,40 €
    13,40 €
    1 * 1 items
       13,40 €
Product Information
  • Colour
    Capacity
    Height
    Availability
    Price
  • Translucent
    14 L
    290 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 33,40 €
    33,40 €
    1 * 1 items
       33,40 €