ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 2.0 N in aqueous solution, Acculute™ standard volumetric solution
Corrosive
Sulphuric acid concentrate for dilution to 2.0 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88568.LQ
CAS Number:  
Corrosive
Sulphuric acid concentrate for dilution to 2.0 N in aqueous solution, Acculute™ standard volumetric solution
Catalog # 88568.LQ
CAS Number:  
Specifications
  • Pk:
    1
  • Seq:
    0001
Specifications
About this item
CAS Number:7664-93-9
 :
Formula:H₂SO₄
MDL Number:MFCD00064589
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS05