Volumetric flasks, USP, class A, blue markings, BLAUBRAND®
Supplier: Brand
Certificates
About this item
Borosilicate glass 3,3 with clear glass stopper or PP stopper.
- Calibrated to contain (TC, In)
- Narrow neck
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- With USP batch certificate or with USP individual certificate
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Specifications
- Catalog No:
- 612-5447
- 612-5451
- 612-5450
- 612-5449
- 612-5432
- 612-5445
- 612-5446
- 612-5438
- 612-5444
- 612-5437
- 612-5443
- 612-5435
- 612-5434
- 612-4870
- 612-5439
- 612-5413
- 612-4813
- 612-4818
- 612-4816
- 612-4821
- 612-5442
- 612-4817
- 612-5433
- 612-4820
- 612-5441
- 612-5431
- 612-5410
- 612-5429
- 612-5440
- 612-5430
- 612-5412
- 612-5411
- 612-5409
- 612-5408
- 612-4814
- 612-5416
- 612-5415
- 612-4815
- 612-5414
- 612-4822
- 612-5448
- 612-4823
- 612-5418
- 612-5417
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Specifications
Related Information
Certificates
Search by Lot NumberBrand BLAUBRAND® Volumetric Instruments USP Brand Volumetric Measurement in the Laboratory
Product Literature
Technical Note BRAND Volumetric Flasks
Application Notes
Frequently Bought Together