Orders LinkContactUsLinkComponent
Forceps holder
Forceps holder
Forceps holder, 180×60×135 mm
Catalog # 232-0189
Supplier:   RSG
Forceps holder
Catalog # 232-0189
Supplier:  RSG
Forceps holder
Forceps holder
Forceps holder, 180×60×135 mm
Price
116,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Made of stainless steel, ESD coated.
  • For safe storage of all forceps used in the lab
  • Ideal for drying forceps after cleaning
  • Acid resistant, anti-static
  • Can be fastened on tables or under cupboards
Specifications
  • Description:
    Forceps holder
  • W×D×H:
    180×60×135 mm