Orders LinkContactUsLinkComponent
Stand for burettes
Stand for burettes
Baseplate with burette support rod (Lר: 505×12 mm), 210×155 mm, Compact automatic burettes
Catalog # BRND23882
Supplier:   Brand
Stand for burettes
Catalog # BRND23882
Supplier:  Brand
Stand for burettes
Stand for burettes
Baseplate with burette support rod (Lר: 505×12 mm), 210×155 mm, Compact automatic burettes
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PP (baseplate) and stainless steel (support rod).
  • Baseplate (L×W: 210×155 mm) with rubber feet which minimise sliding and protect the bench-top
  • Support rod (Lר: 505×12 mm) made of stainless steel
Specifications
  • For:
    Compact automatic burettes
  • Description:
    Baseplate with burette support rod (Lר: 505×12 mm)
  • L×W:
    210×155 mm