Orders LinkContactUsLinkComponent
Buckets
Buckets
Supplier:   Brand
Buckets
Supplier:  Brand
Certificates
Buckets
2 Options Available Below
Product Details & Documents
PP, translucent.
  • Graduated
  • Very good thermal resistance
  • Very good chemical resistance
Universal multipurpose bucket with spout, reinforced rim and handle.
Product Family Options
Product Information
  • Colour
    Capacity
    Height
    Availability
    Price
  • Transparent
    12 L
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 35,30 €
    35,30 €
    1 * 1 items
       35,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRND72376
  • Transparent
    15 L
    340 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 38,20 €
    38,20 €
    1 * 1 items
       38,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRND72378