Orders LinkContactUsLinkComponent
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™
Containers with optional snap caps, Nunc™
PS, clear.
  • Transparent polystyrene construction
  • Versatile containers made of high-quality resins
  • Snap caps available for all containers
  • Cylindrical shape
  • Non sterile
 : Caps must be ordered seperately.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Catalog No:
  • 391-8712
  • 391-8710
  • 391-8713
  • 391-8580
  • 391-8714
  • 391-8579
  • 391-8581
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
Product Family Options
Containers
  • Capacity
    Height
    Type
    Cap colour
    Packed
  • 12,5 ml
    29 mm
    Non sterile, without cap
    Bag of 200, Carton of 1000
    1 * 1.000 items
    1 * 1.000 items - 227,00 €
    227,00 €
    1 * 1.000 items
       227,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8712
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 20 ml
    41 mm
    Non sterile, without cap
    Bag of 100, Carton of 2400
    1 * 2.400 items
    1 * 2.400 items - 678,00 €
    678,00 €
    1 * 2.400 items
       678,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8710
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 30 ml
    60 mm
    Non sterile, without cap
    Bag of 30, Carton of 1200
    1 * 1.200 items
    1 * 1.200 items - 1 190,00 €
    1 190,00 €
    1 * 1.200 items
       1 190,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8713
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 50 ml
    49 mm
    Non sterile, without cap
    Bag of 40, Carton of 800
    1 * 800 items
    1 * 800 items - 375,00 €
    375,00 €
    1 * 800 items
       375,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8580
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 100 ml
    54 mm
    Non sterile, without cap
    Bag of 300, Carton of 300
    Specifications
    Cat. No.
    391-8714
    Environmentally Preferable
    Environmental certification
Snap caps for containers
  • Capacity
    Height
    Type
    Cap colour
    Packed
  • for 12,5/20 and 30 ml containers
    Natural
    1 * 4.000 items
    1 * 4.000 items - 519,00 €
    519,00 €
    1 * 4.000 items
       519,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8579
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • for 50 ml containers
    Grey
    1 * 2.400 items
    1 * 2.400 items - 520,00 €
    520,00 €
    1 * 2.400 items
       520,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-8581
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact