Order Entry
Slovenia
ContactUsLinkComponent
Organic reference standard, Acenaphthene 5000 µg/ml in methanol
Organic reference standard, Acenaphthene 5000 µg/ml in methanol
  PROOU-EPA-1064
 :  
Organic reference standard, Acenaphthene 5000 µg/ml in methanol
  PROOU-EPA-1064
 :  

 

  • Pk:
    1
  • CAS No.:
    83-32-9
  • Description:
    Acenaphthene 5,000 µg/ml in methanol
  • Formula:
    C₁₂H₁₀
  • Description:
    Acenaphthene