Ethyl-3-phenylpropionate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2021-28-5
:
Formula:C₁₁H₁₄O₂
MDL Number:MFCD00009206
:178.23 g/mol
Boiling Pt:118 °C (3 mmHg)
Density:1.016
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
EINECS:217-966-6
Frequently Bought Together