Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Standards
Organic Standards
Pesticide reference standard, 2,4,6-Trichlorophenol 100 µg/ml in methanol
Pesticide reference standard, 2,4,6-Trichlorophenol 100 µg/ml in methanol
PROOU-PH-200-1
:
LGC Standards PROMOCHEM
:
Pesticide reference standard, 2,4,6-Trichlorophenol 100 µg/ml in methanol
PROOU-PH-200-1
:
LGC Standards PROMOCHEM
:
Pk:
1
CAS No.:
88-06-2
Description:
2,4,6-Trichlorophenol 100 µg/ml in methanol
Formula:
C₆H₃Cl₃O
Description:
2,4,6-Trichlorophenol