Orders LinkContactUsLinkComponent
Labels, polypaper, Nalgene®
Labels, polypaper, Nalgene®
PolyPaper right-to-know custom labelling system, Polyolefin, Multicolour, 127×76 mm
Catalog # NALG6316-1000
   Low Manufacturing Impact
Labels, polypaper, Nalgene®
Catalog # NALG6316-1000
   Low Manufacturing Impact
Labels, polypaper, Nalgene®
Labels, polypaper, Nalgene®
PolyPaper right-to-know custom labelling system, Polyolefin, Multicolour, 127×76 mm
Price
159,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Create labels that identify the chemical and its properties. Refer to the appropriate Material Safety Data Sheet (MSDS) or other reference material to verify the correct codes. Overlay is provided to protect the label from chemicals. Consists of pressure-sensitive stickers and blank labels. Complete instructions included with each package.
Specifications
  • Colour:
    Multicolour
  • Description:
    PolyPaper right-to-know custom labelling system
  • Dimensions:
    127×76 mm
  • Material:
    Polyolefin
  • Cat. No.:
    NALG6316-1000
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact