Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid standardised solution 5.0 N in aqueous solution
Sulphuric acid standardised solution 5.0 N in aqueous solution
Sulphuric acid standardised solution 5.0 N in aqueous solution
35613.K2
:
Thermo Fisher Scientific
:
Sulphuric acid standardised solution 5.0 N in aqueous solution
35613.K2
:
Thermo Fisher Scientific
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
7664-93-9
:
:
H₂SO₄
:
MFCD00064589
:
98.08 g/mol
:
1.146 g/cm³ (20 °C)
:
Ambient
:
231-639-5
:
2796
:
8,II
:
GHS05