Hydrochloric acid 0.5 mol/l (0.5 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Titer (mol/l):0.4975 - 0.5025
- Ammonium (NH4):max. 3
- Iron (Fe):max. 0.1
- Heavy Metals (as Pb):max. 0.5
- Sulfate (SO4):max. 1