Sample dippers
Supplier: Brand
Jump to:
About this item
Sampling dipper with reinforced rim and spout.
- PTFE handle with steel core
Optional extension rod for increased handle length.
4 Options Available Below
Product Details & Documents
Sampling dipper with reinforced rim and spout.
- PTFE handle with steel core
Optional extension rod for increased handle length.
Product Family Options
Product Information
- MaterialLengthØCapacityAvailabilityPrice
- BRND90438Sampling dipper with reinforced rim and spoutSampling dipper with reinforced rim and spoutPTFE/steel65 mm55 mm100 mlLog in for availability1 * 1 items154,00 €
- BRND90448Sampling dipper with reinforced rim and spoutSampling dipper with reinforced rim and spoutPTFE/steel95 mm70 mm250 mlLog in for availability1 * 1 items188,00 €
- BRND90454Sampling dipper with reinforced rim and spoutSampling dipper with reinforced rim and spoutPTFE/steel125 mm80 mm500 mlLog in for availability1 * 1 items218,00 €
- BRND90462Sampling dipper with reinforced rim and spoutSampling dipper with reinforced rim and spoutPTFE/steel155 mm105 mm1000 mlLog in for availability1 * 1 items262,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extension rod for sampling device, 600 mm