ContactUsLinkComponent
Accessories for lab tangential flow filtration devices Ultrasette™
Accessories for lab tangential flow filtration devices Ultrasette™
Catalog # 516-8190
undefined
Accessories for lab tangential flow filtration devices Ultrasette™
Catalog # 516-8190
Specifications
  • Description:
    Gauge fitting package, including 1× 0 - 4,1 bar 3,2 mm NPT pressure gauge, 2× 6,4 mm OD PP barbed tube to 3,2 mm NPT connectors, 1× 3,2 mm threaded PP tee, 1× screw clamp, 2× stainless steel tubing clamps
  • Pk:
    1