Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrochloric acid
Hydrochloric acid 6 mol/l (6 N)
Hydrochloric acid 6 mol/l (6 N)
44921.K2
:
Thermo Fisher Scientific
:
Hydrochloric acid 6 mol/l (6 N)
44921.K2
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
1.06 g/cm³ (20 °C)
:
231-595-7
:
8,II
:
36.46 g/mol
:
1789
:
GHS05
:
GHS07
:
Ambient
:
7647-01-0
:
HCl
:
MFCD00011324