Cleanroom face masks, Kimtech™ M6, PE/PP/BiCoSof™
113-0346
- Type:With earloops
- Colour:White
- Environmentally Preferable:
- Pk:500
This gross particulate mask provides basic protection for low-end cleanrooms. It contains no filter.
- Two-layer design
- BiCoSof™ fabric inner facing provides comfort
- Ultrasonic bonding
Suitable for class 1000, ISO 6, M4.5 cleanrooms.