Order Entry
Slovenia
ContactUsLinkComponent
Praseodymium standard solution, 10,000 mg/L Pr in dil. nitric acid standard for AAS
Praseodymium standard solution, 10,000 mg/L Pr in dil. nitric acid standard for AAS
  PROOU-IAA-059
 :  
Praseodymium standard solution, 10,000 mg/L Pr in dil. nitric acid standard for AAS
  PROOU-IAA-059
 :  

 

  • Concentration:
    10000mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Element:
    Praseodymium
  • Symbol:
    Pr