Order Entry
Slovenia
ContactUsLinkComponent
 
Boron standard solution, 1,000 mg/L B in ammonium hydroxide solution (max. 1%) standard for ICP
  PROOU-ICP-005
 :  
Boron standard solution, 1,000 mg/L B in ammonium hydroxide solution (max. 1%) standard for ICP
  PROOU-ICP-005
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    <1% NH₄OH
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Boron
  • Symbol:
    B