Automatic viscometers, ViscoSystem® AVS® 470
About this item
The ViscoSystem AVS 470 is the first measuring device that allows suction and pressure measurements completely independent of a PC.
- Determination with pressure or with vacuum for high viscosities
- Both detections methods optical or thermal are possible
- Nearly all necessary calculations are integrated
- Additional parameters can be set up over a regular PS 2 keyboard
This brings maximum independence and flexibility, allowing you to set up a measuring station, that meets highest requirements, even under difficult conditions, to monitor production or control quality in the polymer and mineral oil industry.
The ViscoSystem AVS 470 determines the time which the sample to be examined requires to flow through the measuring distance in the capillary viscometer with quartz precision.
All common types of viscosity calculations are already integrated into the device, a small PS 2 keyboard is all you need to enter additional data.
A serial printer can be used to conveniently document your measuring results.
2 Options Available Below
Product Details & Documents
The ViscoSystem AVS 470 is the first measuring device that allows suction and pressure measurements completely independent of a PC.
- Determination with pressure or with vacuum for high viscosities
- Both detections methods optical or thermal are possible
- Nearly all necessary calculations are integrated
- Additional parameters can be set up over a regular PS 2 keyboard
This brings maximum independence and flexibility, allowing you to set up a measuring station, that meets highest requirements, even under difficult conditions, to monitor production or control quality in the polymer and mineral oil industry.
The ViscoSystem AVS 470 determines the time which the sample to be examined requires to flow through the measuring distance in the capillary viscometer with quartz precision.
All common types of viscosity calculations are already integrated into the device, a small PS 2 keyboard is all you need to enter additional data.
A serial printer can be used to conveniently document your measuring results.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 616-0026
- 616-1115
- Measurement range (flow time):
- <9999,99 s
- <9999,99 s
- Pump pressure:
- 160 mbar
- 160 mbar
- Viscosity range:
- Pressure: 0,35 - 1800 mm²/s (cSt)
Suction capacity: 0,35 - ~5000 mm²/s (cSt) - Pressure: 0,35 - 1800 mm²/s (cSt)
Suction capacity: 0,35 - ~5000 mm²/s (cSt)
- Temperature regulation period:
- 0 – 20 minutes
- 0 – 20 minutes
- Preselection of the number of measurements:
- Up to 99
- Up to 99
- Time measurement accuracy:
- ±0,01 %
- ±0,01%
- Display:
- LCD
- LCD
- Interfaces:
- RS232C for serial printer
- RS232C for serial printer
- W×D×H:
- 225×320×205 mm
- 225×320×205 mm
- Weight:
- 5,4 kg
- 5,4 kg
Specifications
Product Family Options
Product Information
- ModelRangeAvailabilityPrice
- 616-0026Automatic viscometer with ViscoPump II module for optoelectronic sensingAutomatic viscometer with ViscoPump II module for optoelectronic sensingAVS 4700,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)Log in for availability1 * 1 items11 060,00 €Specifications:
More
Specifications
Cat. No.616-0026Measurement range (flow time)<9999,99 sPump pressure160 mbarViscosity rangePressure: 0,35 - 1800 mm²/s (cSt)
Suction capacity: 0,35 - ~5000 mm²/s (cSt)Temperature regulation period0 – 20 minutesPreselection of the number of measurementsUp to 99Time measurement accuracy±0,01 %DisplayLCDInterfacesRS232C for serial printerW×D×H225×320×205 mmWeight5,4 kg - 616-1115Automatic viscometer with ViscoPump II module for TC sensingAutomatic viscometer with ViscoPump II module for TC sensingAVS 4700,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)Log in for availability1 * 1 items11 060,00 €Specifications:
More
Specifications
Cat. No.616-1115Measurement range (flow time)<9999,99 sPump pressure160 mbarViscosity rangePressure: 0,35 - 1800 mm²/s (cSt)
Suction capacity: 0,35 - ~5000 mm²/s (cSt)Temperature regulation period0 – 20 minutesPreselection of the number of measurementsUp to 99Time measurement accuracy±0,01%DisplayLCDInterfacesRS232C for serial printerW×D×H225×320×205 mmWeight5,4 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, NBR (Perbunan), VZ 7052
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, FKM (viton), VZ 7050
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, EPDM (buna), VZ 7051
Product Information
- Catalog #AvailabilityPrice
- Safety bottle with glass thread, VZ 7021Safety bottle with glass thread, VZ 7021SCOI285420269Log in for availability1 * 1 items - 133,00 €
Product Information
- Catalog #AvailabilityPrice
- VZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mmVZ 3079 Schlauchkappe, silicon, red, int.Ø 9 mmSCOI285414048Log in for availability1 * 1 items - 55,00 €
Product Information
- Catalog #AvailabilityPrice
- VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 470SCOI285414031Log in for availability1 * 1 items - 343,00 €
Product Information
- Catalog #AvailabilityPrice
- Ink ribbon VZ 6249
Product Information
- Catalog #AvailabilityPrice
- VZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mmVZ 5102 Connection cap 'Vacuum', silicon, red, int.Ø 4,5 mmSCOI285410527Log in for availability1 * 1 items - 40,90 €
Product Information
- Catalog #AvailabilityPrice
- VZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mmVZ 5101 Connection cap 'Venting', silicon, black, int.Ø 4,5 mmSCOI285410519Log in for availability1 * 1 items - 40,90 €
Product Information
- Catalog #AvailabilityPrice
- Thermal paper for Z 900 and Z 910 Z 905
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7053, 8,3×2,4 mm
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7054, 7,0×3,0 mm
Product Information
- Catalog #AvailabilityPrice
- Absorbing filter with activated coal, VZ 7216Absorbing filter with activated coal, VZ 7216SCOI285422215Log in for availability1 * 1 items - 180,00 €
Product Information
- Catalog #AvailabilityPrice
- Connectors, VZ 7211, for absorbing filter VZ 7215, 7216Connectors, VZ 7211, for absorbing filter VZ 7215, 7216SCOI285422167Log in for availability1 * 1 items - 94,00 €
Product Information
- Catalog #AvailabilityPrice
- O-ring seal, VZ 7055, 5,3×1,8 mm
Product Information
- Catalog #AvailabilityPrice
- Absorbing filter, VZ 7215
Product Information
- Catalog #AvailabilityPrice
- Dust protection cover, for thermostatic bathsDust protection cover, for thermostatic bathsSCOI285418573Log in for availability1 * 1 items - 133,00 €