Vacuum safety aspiration system AC 02/AC 04
Supplier: HAEP LABOR
Certificates
About this item
The compact vacuum safety aspiration system, together with the diaphragm pump in stainless steel housing and the attached stainless steel safety bottle stand, forms a complete unit with a low base. Flow rate of the pump: 15 L/min. air, 100 mbar final vacuum.
- System optionally available with thicker, more shock-resistant 2 or 4- litre PP bottle
- Screw cap for the bottles with two quick action couplings that close automatically if the hose connections are disengaged
- Safety filter between the collection bottle and the vacuum pump protects the pump and the surrounding area
- Extra-long (3 m) silicone tubing allows the bottle and the vacuum pump to be placed away from the workbench
- With tube holder for attachment to the workbench for comfortable handling
Frequently Bought Together