Vacuum safety aspiration systems AZ 02/AZ 04
Supplier: HAEP LABOR
Certificates
About this item
Compact vacuum safety aspiration system diaphragm vacuum pump that is ideally suited to the task — vacuums fluid sufficiently quickly and does not process solids when used correctly. Flow rate of the pump: 4 L/min air, 250 mbar final vacuum.
- System optionally available with thicker, more shock-resistant 2-litre or 4-litre PP bottle
- Screw cap for the bottles with two quick action couplings that close automatically if the hose connections are disengaged
- Safety filter between the collection bottle and the vacuum pump protects the pump and the surrounding area
- Extra-long (3 m) silicone tubing allows the bottle and the vacuum pump to be placed away from the workbench
- With tube holder for attachment to the workbench for comfortable handling
Frequently Bought Together