Orders LinkContactUsLinkComponent
Solid sampler, SiloDrill
Solid sampler, SiloDrill
Supplier:   BURKLE
Solid sampler, SiloDrill
Supplier:  BURKLE
Certificates
Solid sampler, SiloDrill
2 Options Available Below
Product Details & Documents
When bulk goods are transported, they may separate out and surface samples may not be representative. The sampler is opened manually once it has reached the required depth.
  • Chamber is opened and closed manually
  • Drill tip for easy boring
  • Using extension rods, samples are taken from depths of up to 3,5 metres
Documents
Product Family Options
Product Information
  • Material
    Length
    Ø
    Volume
    Availability
    Price
  • 300-1019
    Solid sampler
    Solid sampler
    Aluminium
    1500 mm
    90 mm
    400 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 778,00 €
    778,00 €
    1 * 1 items
       778,00 €
  • 300-0108
    Solid sampler
    Solid sampler
    Stainless steel V2A (1.4301)
    1500 mm
    90 mm
    400 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3 750,00 €
    3 750,00 €
    1 * 1 items
       3 750,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extension rod, 1000 mm
    Extension rod, 1000 mm
    300-1020
    Log in for availability
    1 * 1 items - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Transport case
    Transport case
    312-1141
    Log in for availability
    1 * 1 items - 135,00 €