Orders LinkContactUsLinkComponent
VWR®, Sample Dippers
VWR®, Sample Dippers
VWR®, Sample Dippers
Supplier:  VWR Collection
Certificates
VWR®, Sample Dippers
VWR®, Sample Dippers
4 Options Available Below
Product Details & Documents
All sample dippers are made of blue PP and consist of a bottle with snap handle.
  • Containers designed for sample collection and dispatch for testing in the same container, eliminating risk of cross-contamination
  • Sterilised by electron-beam irradiation and individually packed in sealed zip bag to guarantee integrity of sample
Ordering information: Available with either hinged plug or screw cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Colour
    Material
    Length
    Ø
    Capacity
    Closure type
    Availability
    Price
  • Sample dippers
    Blue
    PP
    305 mm
    52 mm
    180 ml
    Screw cap
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 283,00 €
    283,00 €
    1 * 100 items
       283,00 €
  • Sample dippers
    Blue
    PP
    275 mm
    52 mm
    125 ml
    Screw cap
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 332,00 €
    332,00 €
    1 * 100 items
       332,00 €
  • Sample dippers
    Blue
    PP
    275 mm
    43 mm
    90 ml
    Hinged plug
    Log in for availability
    Log in for availability
    1 * 150 items
    1 * 150 items - 435,00 €
    435,00 €
    1 * 150 items
       435,00 €
  • Sample dippers
    Blue
    PP
    275 mm
    30 mm
    40 ml
    Screw cap
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - 629,00 €
    629,00 €
    1 * 250 items
       629,00 €