ContactUsLinkComponent
Repetitive pipette, HandyStep® S
  612-3858
 :  Brand
undefined
Repetitive pipette, HandyStep® S
  612-3858
 :  Brand
  • Description:
    HandyStep® S
  • Pk:
    1
Ergonomic and durable repetitive pipette that allows quick and easy serial pipetting and, in conjunction with Brand PD-Tips (10 different sizes from 0,1 µl to 50 ml), providing up to 49 repetitive dispensings from a single aspiration. Also compatible with Combitips®, Combitips® plus and other standard repetitive tips.
  • Easy tip mounting - tips are simply inserted from below
  • Eject key for contact-free, safe tip ejection
  • Superior ergonomic handling
  • DE-M marking, with serial number
  • 59 different volumes with 10 PD-Tip sizes
The HandyStep® S works on the positive displacement principle, making it ideal for routine and research applications, as well as for difficult liquids (viscous, high density or high vapour pressure liquids).
* Stroke setting = % of nominal volume
 : Supplied with DE-M marking, performance certificate, operating manual, shelf/rack mount, three PD-Tips (0,1, 1 and 10 ml).