ContactUsLinkComponent
Nucleic acid building blocks
Nucleic acid building blocks
Catalog # 761-0015
Supplier:  3B Scientific
undefined
Nucleic acid building blocks
Catalog # 761-0015
Supplier:  3B Scientific
Specifications
  • Description:
    Nucleic acid blocks
  • Pk:
    1
Specifications
About this item
Coloured units representing phosphoric acids, purines and pyrimidines, for constructing DNA, t-RNA and RNA helices.
Useful for explaining replication and transcription.