Orders LinkContactUsLinkComponent
Immediate comparison software, Trace
Immediate comparison software, Trace
Software, Trace
Catalog # 630-3042
Supplier:   MOTIC
Immediate comparison software, Trace
Catalog # 630-3042
Supplier:  MOTIC
Immediate comparison software, Trace
Immediate comparison software, Trace
Software, Trace
Price
141,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Trace software enables you to connect two or more Moticams or several digital microscopes to your computer to give you live comparative imaging.
  • Transform your microscopes into comparison microscopes and economically enter the area of forensic science teaching
  • More than just image comparison, you can overlay images in real time and retouch them
  • Authenticate handwriting in a few seconds, or determine whether two pieces of fabric belong to the same batch
  • Easy to install and to use, Trace provides a complete platform for evaluating comparative imaging based reports with accurate measurement and editing tools
  • Will work on PC or MAC
Specifications
  • Description:
    Software, Trace