Orders LinkContactUsLinkComponent
Air flow indicator, Flow Check
Air flow indicator, Flow Check
Air flow indicator set, Flow Check
Catalog # 151-0208
Supplier:   Dräger
Air flow indicator, Flow Check
Catalog # 151-0208
Supplier:  Dräger
Air flow indicator, Flow Check
Air flow indicator, Flow Check
Air flow indicator set, Flow Check
Price
659,00 €
Log in for availability
1 * 1 items
Product Details & Documents
A small heating element in the device head heats an alcohol mixture with a high molecular weight and density similar to air.
If the liquid comes into contact with air, it condenses into a type of mist. The source, flow and speed of the slightest air streams can be identified with the naked eye in this way.
Delivery information: Supplied in a black plastic carrying case with a rechargeable battery pack and a pack of three atomisation cartridges. There is also plenty of space in the carrying case for the battery charger, spare battery and the adapter for the in-car cigarette lighter. The charging device must be ordered separately.
Specifications
  • Model:
    Flow Check
  • Description:
    Air flow indicator set
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Flow check ampoules (and nozzle)
    Flow check ampoules (and nozzle)
    151-0117
    Log in for availability
    1 * 3 items - 41,75 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery pack
    Battery pack
    118-0389
    Log in for availability
    1 * 1 items - 118,00 €