ContactUsLinkComponent
Determination of crude cellulose fibre, ADF and NDF, FibreBag
Determination of crude cellulose fibre, ADF and NDF, FibreBag
Catalog # 705-0469
Supplier:  GERHARDT
undefined
Determination of crude cellulose fibre, ADF and NDF, FibreBag
Catalog # 705-0469
Supplier:  GERHARDT
Specifications
  • Array:
    6×6
  • Pk:
    1
  • Type:
    FBS 36
Specifications
About this item
For rapid determination in accordance with Weender, Van Soeste et al. Use fine mesh bags to weigh the sample. A glass diffuser is then placed inside the bag, allowing the reagents to diffuse freely. Six samples can be processed simultaneously: clean, boil in sulphuric acid, rinse, boil in KOH, rinse again and dry.
  • Considerably reduces reagent consumption
  • Complies with official methods
A six-piece pack includes the following equipment: heating plate, six-piece carousel, handle for the carousel, six glass diffusers, plate stand, glass condenser and tubing.
Accessories available on request.