Systematic On-Line analyser, TresCon® Uno
708-0052
- Description:TresCon® Uno basic unit with nitrate module, TCU/N511
- Environmentally Preferable:
- Pk:1
The TresCon® Uno is a single channel analyzer designed for control and monitoring at waste water treatment plants.
- Automatic diagnosis functions help the user with operation and maintenance
- Modular design enables a quick and easy exchange or replacement of modules
- Easy access and maintenance
- Range of analyser modules are available for many parameters including ammonium-nitrogen, nitrate-nitrogen, orthophosphate, total phosphorus
- With RS232 and RS485 interfaces
The compact size of TresCon® Uno at a reasonable price offers to the user a good price-performance-ratio for the measurement of nutrient parameters. The user interface within the system mounting is simple to operate and has a high resolution, backlit, graphics display.
: The TresCon® Uno consists of the basic controller unit, an analyser module, and a reagent tray. The system can be wall mounted.
For the full range of parameter modules and specification please contact the Avantor specialist.
For the full range of parameter modules and specification please contact the Avantor specialist.