ContactUsLinkComponent
Dissolved oxygen meter, bench, 9500
Catalog # 664-0006
Supplier:  Cole-Parmer
undefined
Dissolved oxygen meter, bench, 9500
Catalog # 664-0006
Supplier:  Cole-Parmer
Specifications
  • Temperature range:
    –10,0...+60,0 °C
  • Description:
    Bench dissolved oxygen meter 9500
  • Model:
    9500
  • Temperature accuracy:
    ±0,5 °C
  • Temperature resolution:
    0,1 °C
  • W×D×H:
    210×250×55 mm
  • Compliance:
    GLP
  • DO concentration:
    0 - 199% / 0 - 25,0% / 0,00 - 19,99 mg/l
  • DO concentration resolution:
    1% / 0,1% / 0,01 mg/L
  • DO accuracy:
    ±2% within 10 °C of calibration temperature
  • Data logging capacity:
    250 reading memory results plus 20 B.O.D. tests (up to 10 samples per test)
  • Display:
    Backlit 1/8 VGA monochrome LCD
  • Outputs:
    Analogue 1 mV per digit, RS232 serial and IrDA printer interface, alarm
  • Temperature compensation:
    0 to +60 °C
  • Weight:
    0,850 kg
  • Pk:
    1
Specifications
About this item
Fully specified dissolved oxygen meter from Jenway that includes full support for the B.O.D 5-day test protocol. Remote monitoring and data collection are facilitated by the provision of both analogue and digital outputs and in the event of power removal all user-defined parameters are retained.
  • Choice of display modes; % air saturation or % oxygen or concentration in mg/L
  • Calibration data status display
  • Comprehensive data logging options
  • GLP functions: Calibration reminder interval (1 to 999 hours); alarm outputs (open collector and audible); security code protected user data
  • Power supply: Battery eliminator
Ordering information: Supplied with DO probe with ATC, electrode holder, UK power supply, spare membranes and KCl solution.