behrotest® DET supplementary set for readily liberated cyanides DLFC
Catalog # 705-0025
Supplier: BEHR
Specifications
- Description:DET Supplementary set for Readily Liberated Cyanides
- Environmentally Preferable:
- Pk:1
- Type:DLFC
Specifications
About this item
For the determination of readily liberated cyanides, the DET basic unit (DBAS, 705-0023) and the supplementary set for total cyanides (DGC, 705-0024) are also required together with the DLFC set.
The DLFC set includes the following equipment:
1 pH electrode for the cyanide determination including cable with BNC plug.
5 Round-bottom reaction vessels, 500 ml.