Distillation, extraction and thermal reactions, behrotest® DET systems
Catalog # 705-0023
Supplier: BEHR
Specifications
- Description:DET basic unit
- Environmentally Preferable:
- Pk:1
- Type:DBAS
Specifications
About this item
The behrotest® universal analytical DET system is a flexible modular system designed for laboratories with frequently changing applications who do not want to invest in dedicated complete apparatus for each application. The system has a core or basic unit to which application specific supplementary/expansion sets can be added. The expansion kits comprise typically of the additional specific glassware and gas flow connections.
The behrotest® DET DBAS includes the following equipment: